Please login to your online account to display your discounted pricing

CD15 Mouse anti-Human, APC, Clone: MMA, eBioscience™

Mouse Monoclonal Antibody

$162.40 - $397.30

Specifications

Antigen CD15
Clone MMA
Host Species Mouse
Gene Alias Leu-M1, Lewis-x, Lex, stage-specific embryonic antigen-1
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123037

 
life technologies
LS17015842
100 tests Each for $397.30

501123036

 
life technologies
LS17015841
25 tests Each for $162.40
Description & Specifications

Specifications

Antigen CD15
Clone MMA
Host Species Mouse
Gene Alias Leu-M1, Lewis-x, Lex, stage-specific embryonic antigen-1
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate APC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

The MMA monoclonal antibody reacts with human CD15, Lewis X. This 3-fucosyl-N-acetyllactosamine carbohydrate moiety is expressed by granulocytes. Monocytes express this structure at varying degree while lymphocytes are negative.