Please login to your online account to display your discounted pricing

CD147 Mouse anti-Human, PE, Clone: 8D12, eBioscience™

Mouse Monoclonal Antibody

$143.55 - $346.55

Specifications

Antigen CD147
Clone 8D12
Host Species Murine
Gene Alias Basigin, EMMPRIN
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010640

 
life technologies
LS12147241
25 Tests Each for $143.55

501122275

 
life technologies
LS12147242
100 Tests Each for $346.55
Description & Specifications

Specifications

Antigen CD147
Clone 8D12
Host Species Murine
Gene Alias Basigin, EMMPRIN
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

The monoclonal antibody 8D12 recognizes CD147 which is a member of the immunoglobulin superfamily. It is also called EMMPRIN and basigin. CD147 has been characterized as a tumor cell collagenase stimulating molecule and has also been shown to have a role in cell adhesion. CD147 can be found in association with integrins alpha3 beta1 and alpha6 beta1 by co-immpunopreciptations. Additionally it is thought to play a role in T cell activation and thymocyte cycling. CD147 is expressed broadly on hematopoietic and non-hematopoietic cells, especially in tumor cells.