missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD146 Mouse, eFluor 450, Clone: P1H12, eBioscience™

Mouse Monoclonal Antibody

$146.52 - $368.28

Specifications

Antigen CD146
Clone P1H12
Host Species Murine
Gene Alias Melanoma Cell Adhesion Molecule, MUC18, Mel-CAM, EndoCAM
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124142 Life Technologies
LS48146941
25 Tests Each for $146.52
501124143 Life Technologies
LS48146942
100 Tests Each for $368.28
Specifications

Description

The monoclonal antibody P1H12 recognizes CD146 also known as MUC18, s-endo, Endo-CAM and Mel-CAM, which is a member of the Ig superfamily of proteins. The expression of CD146 is found on endothelial cells, bone marrow fibroblasts and some tumors (especially melanoma). Recently mesenchymal stromal cells and endometrial stromal cells have also been shown to express CD146. The presence of CD146 on circulating blood cells have been confined to a subset of T cells rather than circulating endothelial cells, as expression of other endothelial markers (CD31 and CD51/61) is negative. Expression can be found on activated lymphocytes. The protein is heavily glycosylated with more than 50% of the mass from carbohydrates.

The antibody P1H12 has been reported to crossreact to mouse, rabbit, canine, but not rat.
Description & Specifications

Specifications

CD146
Murine
Flow Cytometry
eFluor 450
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.125μg)/test
P1H12
Melanoma Cell Adhesion Molecule, MUC18, Mel-CAM, EndoCAM
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer