missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD144 (VE-Cadherin) Rat anti-Mouse, PE-Cy7, Clone: eBioBV13 (BV13), eBioscience™

Rat Monoclonal Antibody

$183.48 - $460.68

Specifications

Antigen CD144 (VE-Cadherin)
Clone eBioBV13 (BV13)
Host Species Rat
Gene Alias Cdh5, Cadherin-5, Vascular endothelial cadherin
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123337 Life Technologies
LS25144180
25μg Each for $183.48
501129219 Life Technologies
LS25144182
100μg Each for $460.68
Specifications

Description

The eBioBV13 (BV13) monoclonal antibody reacts with mouse VE-Cadherin (CD144). VE-Cadherin is a 120kDa member of the type II Cadherin family, characterized by the presence of 5 extracellular cadherin domains (ECD), and anchored to the actin cytoskeleton through their cytoplasmic tail. VE-Cadherin mediates homophilic adhesion between neighbouring endothelial cells and is localized within specialized structures at cell-cell contacts, called adherens junctions. VE-Cadherin is expressed constitutively throughout the entire vasculature, and is required for numerous endothelial cell functions including migration, survival, contact-dependent growth inhibition and endothelial cell assembly into tubular structures. Furthermore, it is thought that VE-Cadherin+CD45- cells from the yolk sac or aorta-gonad-mesonephros (AGM) have the potential to give rise to hematopoietic cells.
Description & Specifications

Specifications

CD144 (VE-Cadherin)
Rat
Flow Cytometry
PE-Cy7
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
0.2mg/mL
eBioBV13 (BV13)
Cdh5, Cadherin-5, Vascular endothelial cadherin
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer