Please login to your online account to display your discounted pricing

CD144 (VE-Cadherin) Rat anti-Mouse, eFluor 450, Clone: eBioBV13 (BV13), eBioscience™

Rat Monoclonal Antibody

$201.55 - $404.55

Specifications

Antigen CD144 (VE-Cadherin)
Clone eBioBV13 (BV13)
Host Species Rat
Gene Alias Cdh5, Cadherin-5, Vascular endothelial cadherin
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5016332

 
life technologies
LS48144180
25μg Each for $201.55

5016333

 
life technologies
LS48144182
100μg Each for $404.55
Description & Specifications

Specifications

Antigen CD144 (VE-Cadherin)
Clone eBioBV13 (BV13)
Host Species Rat
Gene Alias Cdh5, Cadherin-5, Vascular endothelial cadherin
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate eFluor 450
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The BV13 monoclonal antibody reacts with mouse VE-Cadherin (CD144). VE-Cadherin is a 120kDa member of the type II Cadherin family, characterized by the presence of 5 extracellular cadherin domains (ECD), and anchored to the actin cytoskeleton through their cytoplasmic tail. VE-Cadherin mediates homophilic adhesion between neighbouring endothelial cells and is localized within specialized structures at cell-cell contacts, called adherens junctions. VE-Cadherin is expressed constitutively throughout the entire vasculature, and is required for numerous endothelial cell functions including migration, survival, contact-dependent growth inhibition and endothelial cell assembly into tubular structures. Furthermore, it is thought that VE-Cadherin+CD45- cells from the yolk sac or aorta-gonad-mesonephros (AGM) have the potential to give rise to hematopoietic cells.