missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD144 (VE-Cadherin) Mouse anti-Human, PE-Cy7, Clone: 16B1, eBioscience™

Mouse Monoclonal Antibody

$157.08 - $394.68

Specifications

Antigen CD144 (VE-Cadherin)
Clone 16B1
Host Species Murine
Gene Alias Cadherin 5
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123338 Life Technologies
LS25144942
100 Tests Each for $394.68
501128634 Life Technologies
LS25144941
25 Tests Each for $157.08
Specifications

Description

The 16B1 antibody reacts with human CD144, also known as VE-cadherin and cadherin-5. The cadherin family of receptors, which are calcium-dependent adhesion molecules, is known to be involved in homophilic cell interactions. VE-cadherin, which is 140 kDa, is localized at the intercellular boundaries of endothelial cells in blood and lymphatic vessels in several tissues. It is thought to play a role in vascular permeability and remodeling.
Description & Specifications

Specifications

CD144 (VE-Cadherin)
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
16B1
Cadherin 5
Flow Cytometry
PE-Cy7
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
5μL (0.5μg)/test