Please login to your online account to display your discounted pricing

CD144 (VE-Cadherin) Mouse anti-Human, PE, Clone: 16B1, eBioscience™

Mouse Monoclonal Antibody

$172.55 - $448.05

Specifications

Antigen CD144 (VE-Cadherin)
Clone 16B1
Host Species Murine
Gene Alias Cadherin 5
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010635

 
life technologies
LS12144980
25μg Each for $172.55

5010636

 
life technologies
LS12144982
100μg Each for $448.05
Description & Specifications

Specifications

Antigen CD144 (VE-Cadherin)
Clone 16B1
Host Species Murine
Gene Alias Cadherin 5
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The 16B1 antibody reacts with human CD144, also known as VE-cadherin and cadherin-5. The cadherin family of receptors, which are calcium-dependent adhesion molecules, is known to be involved in homophilic cell interactions. VE-cadherin, which is 140 kDa, is localized at the intercellular boundaries of endothelial cells in blood and lymphatic vessels in several tissues. It is thought to play a role in vascular permeability and remodeling.