Please login to your online account to display your discounted pricing

CD144 (VE-Cadherin) Mouse anti-Human, Biotin, Clone: 16B1, eBioscience™

Mouse Monoclonal Antibody

$143.55 - $317.55

Specifications

Antigen CD144 (VE-Cadherin)
Clone 16B1
Host Species Murine
Gene Alias VECadherin, Cadherin 5, Cadherin5
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5011768

 
life technologies
LS13144980
25μg Each for $143.55

5011769

 
life technologies
LS13144982
100μg Each for $317.55
Description & Specifications

Specifications

Antigen CD144 (VE-Cadherin)
Clone 16B1
Host Species Murine
Gene Alias VECadherin, Cadherin 5, Cadherin5
Species Reactivity Human
Applications Flow Cytometry
Applications Immunocytochemistry
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The 16B1 antibody reacts with human CD144, also known as VE-cadherin and cadherin-5. The cadherin family of receptors, which are calcium-dependent adhesion molecules, is known to be involved in homophilic cell interactions. VE-cadherin, which is 140 kDa, is localized at the intercellular boundaries of endothelial cells in blood and lymphatic vessels in several tissues. It is thought to play a role in vascular permeability and remodeling.