Please login to your online account to display your discounted pricing

CD142 Mouse anti-Human, PE, Clone: HTF-1, eBioscience™

Mouse Monoclonal Antibody

$143.55 - $346.55

Specifications

Antigen CD142
Clone HTF-1
Host Species Mouse
Gene Alias tissue factor
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122271

 
life technologies
LS12142941
25 tests Each for $143.55

501122272

 
life technologies
LS12142942
100 tests Each for $346.55
Description & Specifications

Specifications

Antigen CD142
Clone HTF-1
Host Species Mouse
Gene Alias tissue factor
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

This HTF-1 monoclonal antibody reacts with human CD142, which is also known as Tissue Factor. Expression of this type I transmembrane glycoprotein on endothelial cells, monocytes, macrophages, and platelets can be induced by inflammatory mediators (e.g., LPS, IL-1b, TNFa, PMA, or endotoxin). On the other hand, CD142 is expressed constitutively by some tumor cells (e.g., lung, pancreatic, breast, and colon) and non-immune tissues such as the vasculature, central nervous system, kidney, epithelia, and placenta. Studies have also suggested that CD142 exists as a soluble form that circulates in blood. CD142 initiates blood coagulation by associating with and activating the circulating factors VII and VIIa.

The HTF-1 antibody has been reported to exhibit blocking activity.