missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD142 Mouse anti-Human, APC, Clone: HTF-1, eBioscience™

Mouse Monoclonal Antibody

$130.68 - $315.48

Specifications

Antigen CD142
Clone HTF-1
Host Species Murine
Gene Alias Tissue factor
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50245666 Life Technologies
LS17142941
25 Tests Each for $130.68
50245667 Life Technologies
LS17142942
100 Tests Each for $315.48
Specifications

Description

This HTF-1 monoclonal antibody reacts with human CD142, which is also known as Tissue Factor. Expression of this type I transmembrane glycoprotein on endothelial cells, monocytes, macrophages, and platelets can be induced by inflammatory mediators (e.g., LPS, IL-1b, TNFa, PMA, or endotoxin). On the other hand, CD142 is expressed constitutively by some tumor cells (e.g., lung, pancreatic, breast, and colon) and non-immune tissues such as the vasculature, central nervous system, kidney, epithelia, and placenta. Studies have also suggested that CD142 exists as a soluble form that circulates in blood. CD142 initiates blood coagulation by associating with and activating the circulating factors VII and VIIa.

The HTF-1 antibody has been reported to exhibit blocking activity.
Description & Specifications

Specifications

CD142
Murine
Flow Cytometry
Allophycocyanin
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.06μg)/test
HTF-1
Tissue factor
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer