missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Epredia™ Lab Vision™ CD141/Thrombomodulin Ab-1, Mouse Monoclonal Antibody
Optimize immunohistochemical staining of formalin-fixed tissue using Epredia™ CD141/Thrombomodulin Ab-1, Mouse Monoclonal Antibody.
$956.13 - $1543.35
Specifications
Antigen | CD141 |
---|---|
Clone | 141C01; same as 1009 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Thrombomodulin (TM) is also known as Fetomodulin (FM), endothelial anticoagulant protein, or glycoprotein P112. Thrombomodulin contains six repeated domains homologous with epidermal growth factor (EGF) and an amino terminal domain homologous to lectin-like protein. Through its accelerated activation of protein C, synthesis of TM is one of several mechanisms important in maintaining thrombo-resistance and thus reducing clot formation on the surface of the endothelial cells. Immunohistochemical staining of formalin-fixed tissue for TM has been used for the study of a variety of vascular tumors, meningiomas and choriocarcinomas.Host Species: Mouse
Clone: 141C01; same as 1009
Isotype: IgG1/κ
Species Reactivity: Human and Rat. Others not known.
Epitope: EGF domain
Immunogen: Recombinant protein encoding the six repeated EGF domains of human thrombomodulin
Molecular Weight: 75kDa
Positive Control: Mesothelioma
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
CD141 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Rat | |
IgG2a κ |
141C01; same as 1009 | |
Monoclonal | |
Mouse | |
Hematopoietic | |
Recombinant protein encoding the six repeated EGF domains of human thrombomodulin | |
Primary |