missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD141 Rat anti-Mouse, Clone: LS17-9, eBioscience™

Rat Monoclonal Antibody

$146.45 - $353.80

Specifications

Antigen CD141
Clone LS17-9
Host Species Rat
Gene Alias Thrombomodulin, BDCA-3
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122657 Life Technologies
LS14141180
25μg Each for $146.45
501122658 Life Technologies
LS14141182
100μg Each for $353.80
Specifications

Description

The monoclonal antibody LS17-9 recognizes mouse CD141 (Thrombomodulin, BDCA-3), a type I cell surface glycoprotein that binds to and allosterically activates thrombin. This complex cleaves protein C and the Thrombin Activatable Fibrinolysis Inhibitor (TAFI), initiating an important anticoagulant and antifibrinolytic pathway. Although expressed across many tissues and cell types, its highest expression has been reported on the lumenal surface of vascular endothelial cell. In the immune system the highest expression can be observed on monocytes and macrophages. Soluble CD141 is constitutively present in the serum at a very low level, and can be upregulated during inflammation. The anti-inflammatory activity of CD141 results from its ability to block the highly pro-inflammatory activity of thrombin. CD141 prevents chemotaxis and binding of granulocytes to activated endothelium, and inhibits activation of eosinophils and mast cells thereby reducing inflammation. Although human CD141 is commonly used as a marker of cross-presenting dendritic cells, mouse CD141 does not exhibit the same expression pattern.
Description & Specifications

Specifications

CD141
Rat
Mouse
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
LS17-9
Thrombomodulin, BDCA-3
Immunocytochemistry,Immunohistochemistry (Frozen)
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL