missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD140b (PDGF Receptor b) Rat anti-Mouse, Clone: APB5, eBioscience™

Rat Monoclonal Antibody

$71.05 - $143.55

Specifications

Antigen CD140b (PDGF Receptor b)
Clone APB5
Host Species Rat
Gene Alias PDGFRbeta, PDGFR-beta, PDGFR beta, PDGFR b, PDGFR-b
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122656 Life Technologies
LS14140282
100μg Each for $143.55
5012887 Life Technologies
LS14140281
50μg Each for $71.05
Specifications

Description

The APB5 monoclonal antibody reacts with the mouse CD140b molecule, the beta chain of the platelet derived growth factor receptor (PDGF receptor). PDGFRb is a receptor tyrosine kinase that forms dimers on the surface upon ligand binding and phosphorylates substrates. Dimers of PDGFR consist of either homodimers of alpha/alpha, beta/beta, or heterodimers of alpha/beta and serve as a substrate for its kinase activity. CD140b is expressed by embryonic tissues and mesenchymal-derived cells of the adult mouse tissues. The PDGFR beta chain is reported to play a significant role in formation of fibrous atherosclerotic lesions.
Description & Specifications

Specifications

CD140b (PDGF Receptor b)
Rat
Mouse
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
APB5
PDGFRbeta, PDGFR-beta, PDGFR beta, PDGFR b, PDGFR-b
Flow Cytometry,Immunohistochemistry (Frozen)
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL