missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD140a (PDGF Receptor a) Rat anti-Mouse, PE-Cy7, Clone: APA5, eBioscience™

Rat Monoclonal Antibody

$161.04 - $389.40

Specifications

Antigen CD140a (PDGF Receptor a)
Clone APA5
Host Species Rat
Gene Alias Platelet Derived Growth Factor Receptor
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123333 Life Technologies
LS25140180
25μg Each for $161.04
501123334 Life Technologies
LS25140182
100μg Each for $389.40
Specifications

Description

The APA5 monoclonal antibody reacts with the mouse CD140a molecule, the α chain of the platelet derived growth factor receptor (PDGF receptor). PDGFRa is a receptor tyrosine kinase that forms dimers on the surface upon ligand binding and phosphorylates substrates. Dimers of PDGFR consist of either homodimers of α/α, β/β, or heterodimers of α/β and serve as a substrate for its kinase activity. CD140a is expressed by embryonic tissues and mesenchymal-derived cells of the adult mouse tissues.
Description & Specifications

Specifications

CD140a (PDGF Receptor a)
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
APA5
Platelet Derived Growth Factor Receptor
Flow Cytometry
PE-Cy7
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
0.2mg/mL