Please login to your online account to display your discounted pricing

CD14 Rat anti-Mouse, PE, Clone: Sa2-8, eBioscience™

Rat Monoclonal Antibody

$158.05 - $419.05

Specifications

Antigen CD14
Clone Sa2-8
Host Species Rat
Species Reactivity Mouse
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010232

 
affymetrix
12014183
200μg Each for $419.05

501122211

 
affymetrix
12014181
50μg Each for $158.05

501122212

 
affymetrix
12014182
100μg Each for $259.55
Description & Specifications

Specifications

Antigen CD14
Clone Sa2-8
Host Species Rat
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The Sa2-8 monoclonal antibody reacts with mouse CD14, a 53-55kDa GPI-linked glycoprotein. CD14 is a receptor for the complexes of LPS and LBP (LPS-Binding Protein) and is shown to associate with Toll-Like Receptor 4 (TLR4) and participate in the signaling and cellular response to bacterial LPS. In mouse, CD14 is expressed on the surface of macrophages and under certain conditions is also found in the serum in a secreted form. Sa2-8 has weak antagonistic activity (in NF-kappaB activation or TNF alpha production with LPS stimulation).