missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD14 Rat anti-Mouse, Biotin, Clone: Sa2-8, eBioscience™

Rat Monoclonal Antibody

$114.55 - $390.05

Specifications

Antigen CD14
Clone Sa2-8
Host Species Rat
Species Reactivity Mouse
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5011440 Life Technologies
LS13014181
50μg Each for $114.55
5011441 Life Technologies
LS13014182
100μg Each for $216.05
5011442 Life Technologies
LS13014185
500μg Each for $390.05
Specifications

Description

The Sa2-8 monoclonal antibody reacts with mouse CD14, a 53-55kDa GPI-linked glycoprotein. CD14 is a receptor for the complexes of LPS and LBP (LPS-Binding Protein) and is shown to associate with Toll-Like Receptor 4 (TLR4) and participate in the signaling and cellular response to bacterial LPS. In mouse, CD14 is expressed on the surface of macrophages and under certain conditions is also found in the serum in a secreted form. Sa2-8 has weak antagonistic activity (in NF-kappaB activation or TNF alpha production with LPS stimulation).
Description & Specifications

Specifications

CD14
Rat
Flow Cytometry
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL
Sa2-8
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer