Please login to your online account to display your discounted pricing

CD14 Mouse anti-Human, PerCP-eFluor 710, Clone: 61D3, eBioscience™

Mouse Monoclonal Antibody

$234.90 - $516.20

Specifications

Antigen CD14
Clone 61D3
Host Species Murine
Gene Alias LPS-R, Mo2
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5015921

 
life technologies
LS46014941
25 Tests Each for $234.90

5015922

 
life technologies
LS46014942
100 Tests Each for $516.20
Description & Specifications

Specifications

Antigen CD14
Clone 61D3
Host Species Murine
Gene Alias LPS-R, Mo2
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, contains stabilizer if necessary
Concentration 5μL (0.125μg)/test

The 61D3 monoclonal antibody reacts with human CD14, a 53-55kDa GPI-linked glycoprotein. CD14 is expressed on monocytes, interfollicular macrophages and some dendritic cells. Complexes of LPS and LBP (LPS-Binding Protein) bind with high affinity to monocytes through the surface CD14.