Learn More
CD14 Mouse anti-Human, PE, Clone: 61D3, eBioscience™
Mouse Monoclonal Antibody
Manufacturer: Life Technologies LS12014942
Description
Description: The 61D3 monoclonal antibody reacts with human CD14, a 53-55 kDa GPI-linked glycoprotein. CD14 is expressed on monocytes, interfollicular macrophages and some dendritic cells. Complexes of LPS and LBP (LPS-Binding Protein) bind with high affinity to monocytes through the surface CD14. Applications Reported: The 61D3 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 61D3 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD14 is a 55 kDa GPI-anchored glycoprotein that is constitutively expressed on the surface of mature monocytes, macrophages, and neutrophils. CD14 also serves as a multifunctional lipopolysaccharide receptor, and is released to the serum both as a secreted and enzymatically cleaved GPI-anchored form. CD14 binds lipopolysaccharide molecule in a reaction catalyzed by lipopolysaccharide-binding protein (LBP), an acute phase serum protein. The soluble sCD14 can discriminate slight structural differences between lipopolysaccharides and is important for neutralization of serum allochthonous lipopolysaccharides by reconstituted lipoprotein particles. Further, CD14 has been shown to bind apoptotic cells, and can affect allergic, inflammatory and infectious processes. Alternative splicing results in multiple transcript variants encoding the same CD14 isoform. Diseases associated with CD14 dysfunction include mycobacterium chelonae infection and Croup.Specifications
CD14 | |
Monoclonal | |
5 μL/Test | |
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
P08571 | |
monocyte differentiation antigen CD14, myeloid cell-specific leucine-rich glycoprotein | |
Mouse | |
Affinity chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Flow Cytometry | |
61D3 | |
PE | |
CD14 | |
Liquid | |
CD14 | |
IgG1, kappa | |
100 Tests | |
4° C, store in dark, DO NOT FREEZE! | |
Primary | |
929 |
For Research Use Only.