Please login to your online account to display your discounted pricing

CD14 Mouse anti-Human, PE, Clone: 61D3, eBioscience™

Mouse Monoclonal Antibody

$158.05 - $346.55

Specifications

Antigen CD14
Clone 61D3
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010233

 
affymetrix
12014941
25 tests Each for $158.05

501129568

 
affymetrix
12014942
100 tests Each for $346.55
Description & Specifications

Specifications

Antigen CD14
Clone 61D3
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.5μg)/test

The 61D3 monoclonal antibody reacts with human CD14, a 53-55kDa GPI-linked glycoprotein. CD14 is expressed on monocytes, interfollicular macrophages and some dendritic cells. Complexes of LPS and LBP (LPS-Binding Protein) bind with high affinity to monocytes through the surface CD14.