missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD14 Mouse anti-Human, eVolve 605, Clone: 61D3, eBioscience™

Mouse Monoclonal Antibody

$209.88 - $389.40

Specifications

Antigen CD14
Clone 61D3
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501125005 Life Technologies
LS83014941
25 Tests Each for $209.88
501125006 Life Technologies
LS83014942
100 Tests Each for $389.40
Specifications

Description

The 61D3 monoclonal antibody reacts with human CD14, a 53-55kDa GPI-linked glycoprotein. CD14 is expressed on monocytes, interfollicular macrophages and some dendritic cells. Complexes of LPS and LBP (LPS-Binding Protein) bind with high affinity to monocytes through the surface CD14.
Description & Specifications

Specifications

CD14
Murine
Flow Cytometry
eVolve 605
Store at 2-8°C. Do not freeze. Light-sensitive material. This product should be used within 6 months from the date of receipt.
Monoclonal
5μL
61D3
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer