Please login to your online account to display your discounted pricing

CD14 Mouse anti-Human, Biotin, Clone: 61D3, eBioscience™

Mouse Monoclonal Antibody

$129.05 - $274.05

Specifications

Antigen CD14
Clone 61D3
Host Species Mouse
Gene Alias LPS-R, Mo2
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5011443

 
life technologies
LS13014980
25μg Each for $129.05

501128701

 
life technologies
LS13014982
100μg Each for $274.05
Description & Specifications

Specifications

Antigen CD14
Clone 61D3
Host Species Mouse
Gene Alias LPS-R, Mo2
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The 61D3 monoclonal antibody reacts with human CD14, a 53-55kDa GPI-linked glycoprotein. CD14 is expressed on monocytes, interfollicular macrophages and some dendritic cells. Complexes of LPS and LBP (LPS-Binding Protein) bind with high affinity to monocytes through the surface CD14.