Please login to your online account to display your discounted pricing

CD14 Mouse anti-Human, APC-eFluor 780, Clone: 61D3, eBioscience™

Mouse Monoclonal Antibody

$187.05 - $433.55

Specifications

Antigen CD14
Clone 61D3
Host Species Murine
Gene Alias LPS-R, Mo2
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5016147

 
life technologies
LS47014941
25 Tests Each for $187.05

5016148

 
life technologies
LS47014942
100 Tests Each for $433.55
Description & Specifications

Specifications

Antigen CD14
Clone 61D3
Host Species Murine
Gene Alias LPS-R, Mo2
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate APC-eFluor 780
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.5μg)/test

The 61D3 monoclonal antibody reacts with human CD14, a 53-55kDa GPI-linked glycoprotein. CD14 is expressed on monocytes, interfollicular macrophages and some dendritic cells. Complexes of LPS and LBP (LPS-Binding Protein) bind with high affinity to monocytes through the surface CD14.