Please login to your online account to display your discounted pricing

CD138 (Syndecan-1) Mouse anti-Human, PE-Cy7, Clone: DL-101, eBioscience™

Mouse Monoclonal Antibody

$263.90 - $590.15

Specifications

Antigen CD138 (Syndecan-1)
Clone DL-101
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123332

 
life technologies
LS25138941
25 tests Each for $263.90

501129738

 
life technologies
LS25138942
100 tests Each for $590.15
Description & Specifications

Specifications

Antigen CD138 (Syndecan-1)
Clone DL-101
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy7
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.5μg)/test

The DL-101 monoclonal antibody reacts with human CD138, also known as syndecan-1. CD138 is a transmembrane protein containing chondroitin sulfate and heparin sulfate moieties responsible for binding to the extracellular matrix components. CD138 is not expressed by mature B cells but is present on pre-B cells and the finally differentiated plasma cells.