Please login to your online account to display your discounted pricing

CD138 (Syndecan-1) Mouse anti-Human, Clone: DL-101, eBioscience™

Mouse Monoclonal Antibody

$100.05 - $172.55

Specifications

Antigen CD138 (Syndecan-1)
Clone DL-101
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5012883

 
affymetrix
14138980
25μg Each for $100.05

501129463

 
affymetrix
14138982
100μg Each for $172.55
Description & Specifications

Specifications

Antigen CD138 (Syndecan-1)
Clone DL-101
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The DL-101 monoclonal antibody reacts with human CD138, also known as syndecan-1. CD138 is a transmembrane protein containing chondroitin sulfate and heparin sulfate moieties responsible for binding to the extracellular matrix components. CD138 is not expressed by mature B cells but is present on pre-B cells and the finally differentiated plasma cells.