missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD138, Rabbit Polyclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry procedures with Thermo Scientific™ CD138, Rabbit Polyclonal Antibody.

$275.63 - $1,087.22

Specifications

Antigen CD138
Concentration 200µg/mL
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
RB9422P Thermo Scientific™ Lab Vision™
RB9422P
1mL Each for $1,087.22
RB9422P0 Thermo Scientific™ Lab Vision™
RB9422P0
100μL Each for $275.63
RB9422P1 Thermo Scientific™ Lab Vision™
RB9422P1
500μL Each for $770.47
Specifications

Description

Ensure accurate, reproducible results in immunohistochemistry procedures with Thermo Scientific™ CD138, Rabbit Polyclonal Antibody.

CD138/Syndecan-1 is a transmembrane heparin sulphate proteoglycan which is made up of one core protein and five glycosaminoglycan. CD138 is expected to play a role in cell adhesion. It is expressed on the surface of pre-B cells and plasma cells but is absent from mature B cells. It is a selective marker for B cell lymphoblastic leukemia and lymphoplasmocytoid leukemia. It is lost from the apoptotic myeloma cells; hence is a useful marker for viable myeloma cells.

Host Species: Rabbit

Species Reactivity: Human. Others not tested

Epitope: aa 18-218

Immunogen: Recombinant protein encoding aa 18-218 human CD138

Molecular Weight: 30.5kDa (core protein); 85-92kDa (depending upon glycosaminoglycans)

Positive Control: Tonsil

Cellular Localization: Plasma cell and Membrane

Recommended for:

  • Immunohistochemistry

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

CD138
Immunohistochemistry (Paraffin)
Unlabeled
Rabbit
Cell Adhesion
Recombinant protein encoding aa 18-218 human CD138
200µg/mL
Polyclonal
Concentrated
RUO
Primary