missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD137 Ligand (4-1BB Ligand) Rat anti-Mouse, PE, Clone: TKS-1, eBioscience™

Rat Monoclonal Antibody

$143.55 - $433.55

Specifications

Antigen CD137 Ligand (4-1BB Ligand)
Clone TKS-1
Host Species Rat
Gene Alias 4-1BBL, TNFSF9
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5010982 Life Technologies
LS12590181
50μg Each for $143.55
5010984 Life Technologies
LS12590183
200μg Each for $433.55
5010983 Life Technologies
LS12590182
100μg Each for $288.55
Specifications

Description

The TKS-1 monoclonal antibody reacts with mouse 4-1BB Ligand (4-1BBL), a type II transmembrane protein and a member of the tumor necrosis factor superfamily. 4-1BBL is expressed by activated antigen presenting cells including B cells and macrophages. Interaction of 4-1BBL with 4-1BB (CDw137) is reported to deliver a costimulatory signal important in the T-APC interaction.
Description & Specifications

Specifications

CD137 Ligand (4-1BB Ligand)
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
TKS-1
4-1BBL, TNFSF9
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL