missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD137 Ligand (4-1BB Ligand) Mouse anti-Human, PerCP-eFluor™ 710, Clone: 4H3, eBioscience™ DFS Item

Mouse Monoclonal Antibody

$222.00 - $522.00

Specifications

Antigen CD137 Ligand (4-1BB Ligand)
Clone 4H3
Target Species Human
Conjugate PerCP-eFluor™ 710
Isotype IgG1, kappa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
46590641 View Documents Life Technologies
LS46590641
25 Tests Each for $222.00
 
46590642 View Documents Life Technologies
LS46590642
100 Tests Each for $522.00
 
Description

Description

This 4H3 monoclonal antibody recognizes human CD137 ligand, also known as 4-1BB ligand, TNFSF9, or CDw137L. Applications Reported: This 4H3 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 4H3 antibody has been pre-diluted and tested by flow cytometric analysis of stimulated normal human peripheral blood cells. This may be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PerCP-eFluor™ 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product No. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product No. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation.

Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered. CD137 ligand (CD137L) is a type II membrane protein and part of the TNF superfamily. CD137L is a co-stimulatory molecule that is expressed on antigen presenting cells (DCs, monocytes/macrophages, B cells) and is upregulated upon activation. Its receptor, CD137/4-1BB, is found on a variety of cells, including inflamed endothelial cells, where its expression enhances extravasation of CD137L expressing monocytes. Activated T cells also express CD137, and engagement with CD137L enhances T cell proliferation, IL-2 secretion, survival, and cytotoxic activity. CD137L is also expressed on a majority of B-cell lymphomas, including mantle cell lymphoma, follicular lymphoma, and diffuse large B-cell lymphoma (but it is lacking on Hodgkin lymphoma and T-cell lymphoma). CD137L is also expressed in carcinoma cell lines, and is thought to be involved in T cell-tumor cell interactions.
Specifications

Specifications

CD137 Ligand (4-1BB Ligand)
Human
IgG1, kappa
Monoclonal
Liquid
Antibody
4° C, store in dark, DO NOT FREEZE!
4H3
PerCP-eFluor™ 710
Flow Cytometry
Primary
Monoclonal
0.2%PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
RUO