missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD135 (Flt3) Rat anti-Mouse, APC, Clone: A2F10, eBioscience™

Rat Monoclonal Antibody

$118.80 - $322.08

Specifications

Antigen CD135 (Flt3)
Clone A2F10
Host Species Rat
Gene Alias Flk2, Ly-72
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5015076 Life Technologies
LS17135180
25μg Each for $118.80
5015077 Life Technologies
LS17135182
100μg Each for $322.08
Specifications

Description

The A2F10 monoclonal antibody reacts with mouse CD135, also known as Flk2/Flt3. A member of the tyrosine kinase receptor family, this 135-150kDa molecule is expressed by primitive progenitor cells in fetal liver and adult bone marrow. Two-color staining of adult mouse bone marrow cells with A2F10 and RA3-6B2 (CD45R/B220) or M1/70 (CD11b) reveals Flk-2-positive subpopulations of B cells or myeloid cells, respectively.
Description & Specifications

Specifications

CD135 (Flt3)
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
A2F10
Flk2, Ly-72
Flow Cytometry
Allophycocyanin
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL