Learn More
CD135 (Flt3) Monoclonal Antibody (BV10A4H2), PE, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$380.00 - $380.00
Specifications
Antigen | CD135 (Flt3) |
---|---|
Clone | BV10A4H2 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
5010623
|
Invitrogen
12135742 |
100 Tests |
Each for $380.00
|
|
|||||
Description
Description: The BV10A4H2 monoclonal antibody reacts with Human CD135, also known as Flk2 or Flt3. A member of the tyrosine kinase receptor family, this 135-150 kDa molecule is expressed by multipotential progenitor cells including primitive B cell and myelomonocytic progenitors in fetal liver and adult bone marrow. Applications Reported: The BV10A4H2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This BV10A4H2 antibody has been pre-titrated and tested by flow cytometric analysis of REH cell line. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
FLT3 is a tyrosine protein kinase that is a member of the growth factor receptor tyrosine kinase family. The receptor is activated by binding of the fms-related tyrosine kinase 3 ligand to the extracellular domain, which induces homodimer formation in the plasma membrane leading to autophosphorylation of the receptor. The activated receptor kinase subsequently phosphorylates and activates multiple cytoplasmic effector molecules in pathways involved in apoptosis, proliferation, and differentiation of hematopoietic cells in bone marrow. Mutations that result in the constitutive activation of this receptor result in acute myeloid leukemia and acute lymphoblastic leukemia.Specifications
CD135 (Flt3) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P36888 | |
FLT3 | |
IgG1 | |
Affinity chromatography | |
Antibody |
BV10A4H2 | |
Flow Cytometry | |
PE | |
Mouse | |
Human | |
2322 | |
BV-173 leukemic cell line | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
FLT3 |
For Research Use Only.