Learn More
CD135 (Flt3) Monoclonal Antibody (BV10A4H2), APC, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$437.00 - $437.00
Specifications
Antigen | CD135 (Flt3) |
---|---|
Clone | BV10A4H2 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
5015079
|
Invitrogen
17135742 |
100 Tests |
Each for $437.00
|
|
|||||
Description
Description: The BV10A4H2 monoclonal antibody reacts with human CD135, also known as Flk2/Flt3. A member of the tyrosine kinase receptor family, this 135-150 kDa molecule is expressed by multipotential progenitor cells including primitive B cell and myelomonocytic progenitors in fetal liver and adult bone marrow. Applications Reported: The BV10A4H2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This BV10A4H2 antibody has been pre-titrated and tested by flow cytometric analysis of REH cell line. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD135 (Flt3) Monoclonal antibody specifically detects CD135 (Flt3) in Human samples. It is validated for Flow Cytometry.Specifications
CD135 (Flt3) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P36888 | |
FLT3 | |
IgG1 | |
Affinity chromatography | |
Antibody |
BV10A4H2 | |
Flow Cytometry | |
APC | |
Mouse | |
Human | |
2322 | |
BV-173 leukemic cell line | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
FLT3 |
For Research Use Only.