missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD134 (OX40) Rat anti-Mouse, PerCP-eFluor 710, Clone: OX-86, eBioscience™

Rat Monoclonal Antibody

$200.64 - $483.12

Specifications

Antigen CD134 (OX40)
Clone OX-86
Host Species Rat
Gene Alias Ly-70, TNFRSF4
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123772 Life Technologies
LS46134180
25μg Each for $200.64
501123773 Life Technologies
LS46134182
100μg Each for $483.12
Specifications

Description

The OX-86 monoclonal antibody reacts with mouse CD134, also known as OX40. A member of the TNF receptor superfamily, CD134 is a 50kDa type I membrane glycoprotein expressed by activated mouse T lymphocytes. Rat CD134 was initially identified as an activation marker only on activated CD4+ T cells. In contrast, mouse CD134 is expressed by both activated CD4+ and CD8+ T cells. The interaction of CD134 with CD252 (OX40 ligand) has been implicated in T cell-dependent humoral responses, regulation of primary T cell expansion, survival of T cells, size of the memory T cell pool, and regulation of tolerance in the CD4+ T cell compartment.
Description & Specifications

Specifications

CD134 (OX40)
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
OX-86
Ly-70, TNFRSF4
Flow Cytometry
PerCP-eFluor 710
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL