Please login to your online account to display your discounted pricing

CD134 (OX40) Rat anti-Mouse, PE-Cy7, Clone: OX-86, eBioscience™

Rat Monoclonal Antibody

$159.50 - $353.80

Specifications

Antigen CD134 (OX40)
Clone OX-86
Host Species Rat
Gene Alias Ly-70, TNFRSF4
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123330

 
life technologies
LS25134180
25μg Each for $159.50

501129179

 
life technologies
LS25134182
100μg Each for $353.80
Description & Specifications

Specifications

Antigen CD134 (OX40)
Clone OX-86
Host Species Rat
Gene Alias Ly-70, TNFRSF4
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy7
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The OX-86 monoclonal antibody reacts with mouse CD134, also known as OX40. A member of the TNF receptor superfamily, CD134 is a 50kDa type I membrane glycoprotein expressed by activated mouse T lymphocytes. Rat CD134 was initially identified as an activation marker only on activated CD4+ T cells. In contrast, mouse CD134 is expressed by both activated CD4+ and CD8+ T cells. The interaction of CD134 with CD252 (OX40 ligand) has been implicated in T cell-dependent humoral responses, regulation of primary T cell expansion, survival of T cells, size of the memory T cell pool, and regulation of tolerance in the CD4+ T cell compartment.