missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD134 (OX40) Monoclonal Antibody (OX-86), PE-Cyanine7, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

Supplier:  Invitrogen 25134182

 View more versions of this product

Catalog No. 501129179



Description

Description

Description: The OX-86 monoclonal antibody reacts with mouse CD134, also known as OX40. A member of the TNF receptor superfamily, CD134 is a 50 kDa type I membrane glycoprotein expressed by activated mouse T lymphocytes. Rat CD134 was initially identified as an activation marker only on activated CD4+ T cells. In contrast, mouse CD134 is expressed by both activated CD4+ and CD8+ T cells. The interaction of CD134 with CD252 (OX40 ligand) has been implicated in T cell-dependent humoral responses, regulation of primary T cell expansion, survival of T cells, size of the memory T cell pool, and regulation of tolerance in the CD4+ T cell compartment. Applications Reported: This OX-86 antibody has been reported for use in flow cytometric analysis. Applications Tested: This OX-86 antibody has been tested by flow cytometric analysis of stimulated mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat.

CD134 (OX40) Monoclonal antibody specifically detects CD134 (OX40) in Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD134 (OX40)
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P47741
Tnfrsf4
Affinity chromatography
RUO
22163
4° C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
OX-86
PE-Cyanine7
Tnfrsf4
ACT35; ACT35 antigen; ATC35 antigen; CD134; CD134 antigen; IMD16; Ly-70; lymphoid activation antigene ACT35; MRC OX40; Ox40; OX40 antigen; OX40 cell surface antigen; OX40 homologue; OX40L receptor; TAX transcriptionally-activated glycoprotein 1 receptor; tax-transcriptionally activated glycoprotein 1; tax-transcriptionally activated glycoprotein 1 receptor; TNF receptor superfamily member 4; Tnfrsf4; tumor necrosis factor (ligand) superfamily member 4; tumor necrosis factor receptor superfamily member 4; tumor necrosis factor receptor superfamily, member 4; tumor necrosis factor superfamily, member 4; Txgp1; Txgp1l
Rat
100 μg
Primary
Mouse
Antibody
IgG1 κ
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only.