missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD134 (OX40) Mouse anti-Human, FITC, Clone: ACT35 (ACT-35), eBioscience™

Mouse Monoclonal Antibody

$130.68 - $275.88

Specifications

Antigen CD134 (OX40)
Clone ACT35 (ACT-35)
Host Species Murine
Gene Alias OX-40, TNFRSF4
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
509761 Life Technologies
LS11134741
25 Tests Each for $130.68
509762 Life Technologies
LS11134742
100 Tests Each for $275.88
Specifications

Description

The ACT35 monoclonal antibody reacts with human CD134, also known as OX-40. A member of the TNF receptor superfamily, CD134/OX-40 is a 50kDa type I membrane glycoprotein expressed by activated T lymphocytes. The interaction of CD134 with OX-40L has been implicated in T cell-dependent humoral response, regulation of primary T cell expansion, survival of T cells, size of the memory T cell pool and regulation of tolerance in the CD4+ T cell compartment.
Description & Specifications

Specifications

CD134 (OX40)
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
ACT35 (ACT-35)
OX-40, TNFRSF4
Flow Cytometry,Immunohistochemistry (Formalin/Paraffin),Immunohistochemistry (Frozen)
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.5μg)/test