Please login to your online account to display your discounted pricing

CD134 (OX40) Mouse anti-Human, APC, Clone: ACT35, eBioscience™

Mouse Monoclonal Antibody

$158.05 - $346.55

Specifications

Antigen CD134 (OX40)
Clone ACT35 (ACT-35)
Host Species Mouse
Gene Alias OX-40, TNFRSF4
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5015074

 
life technologies
LS17134741
25 tests Each for $158.05

5015075

 
life technologies
LS17134742
100 tests Each for $346.55
Description & Specifications

Specifications

Antigen CD134 (OX40)
Clone ACT35 (ACT-35)
Host Species Mouse
Gene Alias OX-40, TNFRSF4
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate APC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

The ACT35 monoclonal antibody reacts with human CD134, also known as OX-40. A member of the TNF receptor superfamily, CD134/OX-40 is a 50kDa type I membrane glycoprotein expressed by activated T lymphocytes. The interaction of CD134 with OX-40L has been implicated in T cell-dependent humoral response, regulation of primary T cell expansion, survival of T cells, size of the memory T cell pool and regulation of tolerance in the CD4+ T cell compartment.