missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD133 (Prominin-1) Rat anti-Canine, Mouse, PE, Clone: 13A4, eBioscience™

Rat Monoclonal Antibody

Manufacturer:  Life Technologies LS12133182

 View more versions of this product

Catalog No. 501128893



Description

Description

Description: The 13A4 monoclonal antibody recognizes mouse Prominin-1 (sometimes also referred to as CD133 and, in the case of the human orthologue, as AC133), a 115-120 kDa pentaspan transmembrane (5-TM) domain glycoprotein. Prominin-1 is expressed on primitive cells such as hematopoietic stem and progenitor cells, neural and endothelial stem cells, retina and retinoblastoma, as well as developing epithelium. To date, the function and ligand of Prominin-1 are unknown. The 13A4 antibody does not cross react with rat, human, chicken, or Drosophila antigen but has been reported to work in canine/dog. Applications Reported: The 13A4 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 13A4 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 1 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.

The CD133 (also named AC133) antigen is a 97 kDa glycoprotein, with 5 transmembrane domains, that shows up as a 120 kDa band on protein gels due to glycosylation. It is a cell surface marker expressed by immature hematopoietic stem cells but not mature blood cells. In addition to hematopoietic stem and progenitor cells, CD133 has been found to be a marker for other stem and progenitor cells including neural and embryonic stem cells. It has been shown to be expressed in cancers, including some leukemias and brain tumours, that may be derived from stem cells. Low level expression could also be detected in the kidney, pancreas, placenta, and fetal liver.
Specifications

Specifications

CD133 (Prominin-1)
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
O54990
AC133, CD133, CORD12, MCDR2, MSTP061, PROML1, RP41, STGD4
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Canine, Mouse
Flow Cytometry
13A4
PE
Prom1
Liquid
Prom1
IgG1, kappa
100 μg
4° C, store in dark, DO NOT FREEZE!
Primary
19126, 488816
Documents

For Research Use Only.