Learn More
CD133 (Prominin-1) Rat anti-Canine, Mouse, Clone: 13A4, eBioscience™
Rat Monoclonal Antibody
Manufacturer: Life Technologies LS14133182
Description
Description: The 13A4 monoclonal antibody recognizes mouse Prominin-1 (sometimes also referred to as CD133 and, in the case of the human orthologue, as AC133), a 115-120 kDa pentaspan transmembrane (5-TM) domain glycoprotein. Prominin-1 is expressed on primitive cells such as hematopoietic stem and progenitor cells, neural and endothelial stem cells, retina and retinoblastoma, as well as developing epithelium. To date, the function and ligand of Prominin-1 are unknown. The 13A4 antibody does not cross react with rat, human, chicken, or Drosophila antigen but has been reported to work in canine/dog. Applications Reported: The 13A4 antibody has been reported for use in flow cytometric analysis (∽0.25-1 µg/million cells), immunoprecipitation (∽10-25 µg/mL), immunoblotting (WB) (∽1-5 µg/mL), and immunohistochemical staining (∽1-10 µg/mL). Applications Tested: The 13A4 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 1 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE.
Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered. The CD133 (also named AC133) antigen is a 97 kDa glycoprotein, with 5 transmembrane domains, that shows up as a 120 kDa band on protein gels due to glycosylation. It is a cell surface marker expressed by immature hematopoietic stem cells but not mature blood cells. In addition to hematopoietic stem and progenitor cells, CD133 has been found to be a marker for other stem and progenitor cells including neural and embryonic stem cells. It has been shown to be expressed in cancers, including some leukemias and brain tumours, that may be derived from stem cells. Low level expression could also be detected in the kidney, pancreas, placenta, and fetal liver.Specifications
CD133 (Prominin-1) | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
O54990 | |
AC133, CD133, CORD12, MCDR2, MSTP061, PROML1, RP41, STGD4 | |
Rat | |
Affinity chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Canine, Mouse |
Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
13A4 | |
Unconjugated | |
Prom1 | |
Liquid | |
Prom1 | |
IgG1, kappa | |
100 μg | |
4° C | |
Primary | |
19126, 488816 |
For Research Use Only.