missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD133 (Prominin-1) Rat anti-Mouse, Biotin, Clone: 13A4, eBioscience™

Rat Monoclonal Antibody

$158.05 - $404.55

Specifications

Antigen CD133 (Prominin-1)
Clone 13A4
Host Species Rat
Species Reactivity Canine, Mouse
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122527 Life Technologies
LS13133180
25μg Each for $158.05
5011752 Life Technologies
LS13133182
100μg Each for $404.55
Specifications

Description

The 13A4 monoclonal antibody recognizes mouse Prominin-1 (sometimes also referred to as CD133 and, in the case of the human orthologue, as AC133), a 115-120kDa pentaspan transmembrane (5-TM) domain glycoprotein. Prominin-1 is expressed on primitive cells such as hematopoietic stem and progenitor cells, neural & endothelial stem cells, retina and retinoblastoma, as well as developing epithelium. To date, the function and ligand of Prominin-1 are unknown. The 13A4 antibody does not cross react with rat, human, chicken, or Drosophila antigen but has been reported to work in canine/dog.
Description & Specifications

Specifications

CD133 (Prominin-1)
Rat
Flow Cytometry
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL
13A4
Canine, Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer