Please login to your online account to display your discounted pricing

CD133 Mouse anti-Human, PE, Clone: TMP4, eBioscience™

Mouse Monoclonal Antibody

$191.40 - $456.75

Specifications

Antigen CD133
Clone TMP4
Host Species Mouse
Gene Alias AC133, Prominin-1
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122267

 
life technologies
LS12133841
25 tests Each for $191.40

501122268

 
life technologies
LS12133842
100 tests Each for $456.75
Description & Specifications

Specifications

Antigen CD133
Clone TMP4
Host Species Mouse
Gene Alias AC133, Prominin-1
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

The TMP4 monoclonal antibody reacts with human CD133 (Prominin-1), a 120kDa member of the pentaspan family of proteins, which also includes Prominin-2. Their expression is found within plasma membrane protrusions such as epithelial microvilli. CD133 can exist in a number of alternatively spliced isoforms, and the protein has several N-linked glycosylation sites: the occurrence of both may be tissue-dependent. Human CD133 was first identified as an epitope expressed on CD34+ hematopoietic progenitors. Although the ligand and function of CD133 remain unknown, it has since proven to be very useful as a marker for both stem cells and cancer stem cells. In addition to its expression on hematopoietic precursors, CD133 has been used to identify tumorigenic colon cancer stem cells, brain cancer stem cells, prostate cancer stem cells, in addition to others.

The binding of the TMP4 antibody does not block the binding of another anti-human CD133 antibody, EMK08 (cat. 12-1339) indicating that they recognize distinct epitopes.