missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD131 Mouse anti-Human, PE, Clone: 1C1, eBioscience™

Mouse Monoclonal Antibody

$143.88 - $262.68

Specifications

Antigen CD131
Clone 1C1
Host Species Murine
Gene Alias common beta subunit, Colony Stimulating Factor 2 Receptor beta
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50245286 Life Technologies
LS12131941
25 Tests Each for $143.88
50245287 Life Technologies
LS12131942
100 Tests Each for $262.68
Specifications

Description

The 1C1 monoclonal antibody reacts with the human CD131 molecule, also known as the common β subunit (βC). The common β subunit associates with the specific α subunits of IL-3 receptor, IL-5 receptor and GM-CSF receptor to form high affinity receptors for these cytokines. These cytokine receptors are expressed by neutrophils, eosinophils, monocytes, endothelial cells, fibroblasts and hematopoietic progenitor cells and play a crucial role in growth/activation of eosinophils and in the inflammatory response.
Description & Specifications

Specifications

CD131
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
1C1
common beta subunit, Colony Stimulating Factor 2 Receptor beta
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.25μg)/test