Please login to your online account to display your discounted pricing

CD131 Mouse anti-Human, Biotin, Clone: 1C1, eBioscience™

Mouse Monoclonal Antibody

$56.55 - $143.55

Specifications

Antigen CD131
Clone 1C1
Host Species Mouse
Gene Alias common beta subunit, Colony Stimulating Factor 2 Receptor beta
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122525

 
life technologies
LS13131980
25μg Each for $56.55

501122526

 
life technologies
LS13131982
100μg Each for $143.55
Description & Specifications

Specifications

Antigen CD131
Clone 1C1
Host Species Mouse
Gene Alias common beta subunit, Colony Stimulating Factor 2 Receptor beta
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The 1C1 monoclonal antibody reacts with the human CD131 molecule, also known as the common β subunit (βC). The common β subunit associates with the specific α subunits of IL-3 receptor, IL-5 receptor and GM-CSF receptor to form high affinity receptors for these cytokines. These cytokine receptors are expressed by neutrophils, eosinophils, monocytes, endothelial cells, fibroblasts and hematopoietic progenitor cells and play a crucial role in growth/activation of eosinophils and in the inflammatory response.