missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD130 (gp130) Rat anti-Mouse, PE, Clone: KGP130, eBioscience™

Rat Monoclonal Antibody

$130.68 - $315.48

Specifications

Antigen CD130 (gp130)
Clone KGP130
Host Species Rat
Gene Alias gp130, IL-6 Receptor beta, IL-6RB
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122264 Life Technologies
LS12130280
25μg Each for $130.68
501122265 Life Technologies
LS12130282
100μg Each for $315.48
Specifications

Description

The KGP130 monoclonal antibody reacts with mouse gp130, also known as CD130. gp130 is a 130 kD Type-I transmembrane glycoprotein containing a 597 amino acid extracellular domain, a single transmembrane domain, and a 277 amino acid cytoplasmic domain. gp130 is a subunit of several heterodimeric cell-surface receptors, including receptors for IL-6, IL-11, IL-27, Oncostatin M, and Leukemia Inhibitory Factor (LIF). The gp130 protein has also been found to exist in a soluble form, which is capable of inhibiting IL-6 signaling. gp130 is expressed mainly on T cells, monocytes, endothelial cells, activated B cells, and plasma cells, and is expressed at lower levels on most leukocytes and epithelial cells. In response to ligand binding, gp130 becomes tyrosine phosphorylated, leading to activation of several signaling pathways including the PI3 kinase, Ras-MAPk and Stat pathways.

Preincubation of mouse splenic T cells with IL-6 is capable of blocking the interaction of KGP130 with gp130, suggesting that the KGP130 binding site lies near the IL-6 interaction domain. Reactivity of KGP130 towards human gp130 has not been observed.
Description & Specifications

Specifications

CD130 (gp130)
Rat
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
KGP130
gp130, IL-6 Receptor beta, IL-6RB
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer