Please login to your online account to display your discounted pricing

CD13 Mouse anti-Human, PerCP-eFluor 710, Clone: WM-15 (WM15), eBioscience™

Mouse Monoclonal Antibody

$187.05 - $433.55

Specifications

Antigen CD13
Clone WM-15 (WM15)
Host Species Mouse
Gene Alias Alanyl Aminopeptidase (ANPEP), Aminopeptidase N (PEPN)
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5015919

 
life technologies
LS46013841
25 tests Each for $187.05

501130738

 
life technologies
LS46013842
100 tests Each for $433.55
Description & Specifications

Specifications

Antigen CD13
Clone WM-15 (WM15)
Host Species Mouse
Gene Alias Alanyl Aminopeptidase (ANPEP), Aminopeptidase N (PEPN)
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

The WM-15 monoclonal antibody reacts with human CD13, also known as aminopeptidase N. CD13 is a transmembrane ectoenzyme occurring on monocytes and granulocytes but not lymphocytes in blood. It is also reportedly on a wide variety of cells and in a wide variety of tissues, including endothelial cells, epithelial cells, fibroblasts, kidney proximal tubules, intestine, and placenta. CD13 is reported to have many different functions including degradation of enkephalins and endorphins, terminal degradation of peptides, amino acid scavenging, antigen processing, and adhesion and migration of cells.