missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD13 Mouse anti-Human, Clone: WM-15 (WM15), eBioscience™

Mouse Monoclonal Antibody

$64.68 - $130.68

Specifications

Antigen CD13
Clone WM-15 (WM15)
Host Species Murine
Gene Alias Alanyl Aminopeptidase (ANPEP), Aminopeptidase N (PEPN)
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012429 Life Technologies
LS14013880
25μg Each for $64.68
5012430 Life Technologies
LS14013882
100μg Each for $130.68
Specifications

Description

The WM-15 monoclonal antibody reacts with human CD13, also known as aminopeptidase N. CD13 is a transmembrane ectoenzyme occurring on monocytes and granulocytes but not lymphocytes in blood. It is also reportedly on a wide variety of cells and in a wide variety of tissues, including endothelial cells, epithelial cells, fibroblasts, kidney proximal tubules, intestine, and placenta. CD13 is reported to have many different functions including degradation of enkephalins and endorphins, terminal degradation of peptides, amino acid scavenging, antigen processing, and adhesion and migration of cells.
Description & Specifications

Specifications

CD13
Murine
Human
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
WM-15 (WM15)
Alanyl Aminopeptidase (ANPEP), Aminopeptidase N (PEPN)
Flow Cytometry,Immunohistochemistry
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL