Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD13/Aminopeptidase-N Ab-3, Mouse Monoclonal Antibody

Stain granulocytes, monocytes, mast cells, GM-progenitor cells and more with Thermo Scientific™ CD13/Aminopeptidase-N Ab-3, Mouse Monoclonal Antibody.

$368.86 - $1,249.16

Specifications

Antigen CD13/Aminopeptidase-N Ab-3
Clone 38C12
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MS1079R7

 
thermo scientific™ lab vision™
MS1079R7
7mL Each for $926.12

MS1079S

 
thermo scientific™ lab vision™
MS1079S
1mL Each for $1,249.16

MS1079S0

 
thermo scientific™ lab vision™
MS1079S0
100µL Each for $368.86

MS1079S1

 
thermo scientific™ lab vision™
MS1079S1
500µL Each for $840.18
Description & Specifications

Specifications

Antigen CD13/Aminopeptidase-N Ab-3
Clone 38C12
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Mouse
Regulatory Status RUO
Research Discipline Neuroscience
Type Primary
Immunogen Recombinant protein encoding the C-terminal half of the extracellular domain of human CD13
Isotype IgG1

Stain granulocytes, monocytes, mast cells, GM-progenitor cells and more with Thermo Scientific™ CD13/Aminopeptidase-N Ab-3, Mouse Monoclonal Antibody, which provides accurate, reproducible results.

CD13 (also known as aminopeptidase-N) is present on most cells of myeloid origin including granulocytes, monocytes, mast cells, and GM-progenitor cells. It is also expressed by the majority of AML, CML in myeloid blast crisis, and in a smaller fraction of lymphoid leukemias. CD13 is absent from normal lymphocytes, platelets and erythrocytes. CD13 is also present on fibroblasts, endothelial cells, epithelial cells from renal proximal tubules and intestinal brush border, bone marrow stromal cells, osteoclasts, and cells lining bile canaliculi. CD13 plays a role in metabolism of biologically active peptides, in phagocytosis, and in bactericidal/tumoricidal activities. It also serves as a receptor for human coronaviruses (HCV).

Host Species: Mouse

Clone: 38C12

Isotype: IgG1

Species Reactivity: Human. Others not tested.

Epitope: C-terminal half of the extracellular domain

Immunogen: Recombinant protein encoding the C-terminal half of the extracellular domain of human CD13

Molecular Weight: 150kDa

Positive Control: Prostate, liver

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)
  • Western Blotting (Not suitable)

Note:

USA: RUO; Int'l: RUO