Please login to your online account to display your discounted pricing

CD127 Rat anti-Mouse, PerCP-eFluor 710, Clone: eBioSB/199 (SB/199), eBioscience™

Rat Monoclonal Antibody

$158.05 - $419.05

Specifications

Antigen CD127
Clone eBioSB/199 (SB/199)
Host Species Rat
Gene Alias Interleukin-7 Receptor alpha
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123771

 
affymetrix
46127380
25μg Each for $158.05

501129128

 
affymetrix
46127382
100μg Each for $419.05
Description & Specifications

Specifications

Antigen CD127
Clone eBioSB/199 (SB/199)
Host Species Rat
Gene Alias Interleukin-7 Receptor alpha
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The SB/199 monoclonal antibody reacts with mouse CD127, the high affinity a subunit of the mouse IL-7 receptor. IL-7 receptor a chain is expressed by immature B cells in the bone marrow, double-negative (CD4-/CD8-), single-positive (CD4+ and CD8+), but not double-positive (CD4+/CD8+) thymocytes. In the periphery, mature T cells express low levels of CD127. Binding of SB/199 blocks the binding of A7R34, another antibody which recognizes mouse CD127.