Learn More
CD127 Monoclonal Antibody (eBioSB/199 (SB/199)), PE-Cyanine7, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
$182.00 - $478.00
Specifications
Antigen | CD127 |
---|---|
Clone | eBioSB/199 (SB/199) |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
Description: The SB/199 monoclonal antibody reacts with mouse CD127, the high affinity a subunit of the mouse IL-7 receptor. IL-7 receptor a chain is expressed by immature B cells in the bone marrow, double-negative (CD4-/CD8-), single-positive (CD4+ and CD8+), but not double-positive (CD4+/CD8+) thymocytes. In the periphery, mature T cells express low levels of CD127. Binding of SB/199 blocks the binding of A7R34, another antibody which recognizes mouse CD127. Applications Reported: This eBioSB/199 (SB/199) antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBioSB/199 (SB/199) antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat.
CD127 Monoclonal antibody specifically detects CD127 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD127 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
CD127; CD127 antigen; CDW127; IL 7 receptor; IL 7R a; IL 7R subunit alpha; IL 7R a; IL 7Ra; IL 7Ra; IL7 receptor; IL-7 receptor alpha chain; IL7 receptor subunit alpha; IL-7 receptor subunit alpha; IL7R; IL-7R; IL7R a; IL7R alpha; IL7R subunit alpha; IL-7R subunit alpha; IL7R a; IL7RA; IL-7RA; IL-7Ralpha; IL-7R-alpha; IL7Ra; ILRA; interleukin 7 receptor; interleukin 7 receptor alpha chain; interleukin 7 receptor isoform H5-6; Interleukin7 receptor subunit alpha; interleukin-7 receptor subunit alpha; MGC107557; sCD127; soluble IL 7 receptor; soluble IL 7R | |
Il7r | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Il7r |
eBioSB/199 (SB/199) | |
Flow Cytometry | |
PE-Cyanine7 | |
Rat | |
Mouse | |
P16872 | |
16197 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.