missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD127 Rat anti-Mouse, PE, Clone: eBioSB/199 (SB/199), eBioscience™

Rat Monoclonal Antibody

$143.88 - $447.48

Specifications

Antigen CD127
Clone eBioSB/199 (SB/199)
Host Species Rat
Gene Alias Interleukin-7 Receptor alpha
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5010602 Life Technologies
LS12127381
50μg Each for $143.88
5010604 Life Technologies
LS12127383
200μg Each for $447.48
5010603 Life Technologies
LS12127382
100μg Each for $249.48
Specifications

Description

The SB/199 monoclonal antibody reacts with mouse CD127, the high affinity a subunit of the mouse IL-7 receptor. IL-7 receptor a chain is expressed by immature B cells in the bone marrow, double-negative (CD4-/CD8-), single-positive (CD4+ and CD8+), but not double-positive (CD4+/CD8+) thymocytes. In the periphery, mature T cells express low levels of CD127. Binding of SB/199 blocks the binding of A7R34, another antibody which recognizes mouse CD127.
Description & Specifications

Specifications

CD127
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
eBioSB/199 (SB/199)
Interleukin-7 Receptor alpha
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL