missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD127 Rat anti-Mouse, FITC, Clone: A7R34, eBioscience™

Rat Monoclonal Antibody

$118.80 - $469.92

Specifications

Antigen CD127
Clone A7R34
Host Species Rat
Gene Alias Interleukin-7 Receptor alpha, IL-7Ra
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
509752 Life Technologies
LS11127181
50μg Each for $118.80
509753 Life Technologies
LS11127182
100μg Each for $241.56
509754 Life Technologies
LS11127185
500μg Each for $469.92
Specifications

Description

The A7R34 monoclonal antibody reacts with mouse CD127, the high affinity alpha subunit of the mouse IL-7 receptor. IL-7 receptor alpha chain is expressed by immature B cells in the bone marrow, double-negative (CD4-CD8-), single-positive (CD4+ and CD8+), but not double-positive (CD4+CD8+) thymocytes. In the periphery, mature T cells express CD127 at low level. A7R34 inhibits binding of IL-7 to its receptor and has been used in in vivo and in vitro studies to elucidate the role of IL-7 in T and B cell development and activation. Binding of A7R34 blocks the binding of SB/199, another antibody which recognizes mouse CD127.
Description & Specifications

Specifications

CD127
Rat
Flow Cytometry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.5mg/mL
A7R34
Interleukin-7 Receptor alpha, IL-7Ra
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer