Please login to your online account to display your discounted pricing

CD127 Rat anti-Mouse, Alexa Fluor 488, Clone: A7R34, eBioscience™

Rat Monoclonal Antibody

$176.90 - $413.25

Specifications

Antigen CD127
Clone A7R34
Host Species Rat
Gene Alias Interleukin-7 Receptor alpha
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5016726

 
life technologies
LS53127180
25μg Each for $176.90

5016727

 
life technologies
LS53127182
100μg Each for $413.25
Description & Specifications

Specifications

Antigen CD127
Clone A7R34
Host Species Rat
Gene Alias Interleukin-7 Receptor alpha
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Alexa Fluor 488
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The A7R34 monoclonal antibody reacts with mouse CD127, the high affinity alpha subunit of the mouse IL-7 receptor. IL-7 receptor alpha chain is expressed by immature B cells in the bone marrow, double-negative (CD4-CD8-), single-positive (CD4+ and CD8+), but not double-positive (CD4+CD8+) thymocytes. In the periphery, mature T cells express CD127 at low level. A7R34 inhibits binding of IL-7 to its receptor and has been used in in vivo and in vitro studies to elucidate the role of IL-7 in T and B cell development and activation. Binding of A7R34 blocks the binding of SB/199, another antibody which recognizes mouse CD127.